You Searched For: 1-(2-Methylphenyl)urea
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (TCT0802-025G)
Supplier: TCI America
Description: CAS Number: 614-77-7 MDL Number: MFCD00025407 Molecular Formula: C8H10N2O Molecular Weight: 150.18 Purity/Analysis Method: 98.0% (T) Form: Cr...