Phthalamic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Carbamoylbenzoic acid
P0282-25G
TCP0282-025GEA
126.88
USD
TCP0282-025G
Phthalamic acid ≥98.0% (by HPLC, titration analysis)
Phthalamic acid
CAS Number: 88-97-1
MDL Number: MFCD00025476
Molecular Formula: C8H7NO3
Molecular Weight: 165.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 149
Other Product Names / Synonyms: Phthalamidic Acid
Formula:
C₈H₇NO₃ MW: 165.15 g/mol Melting Pt: 149 °C |
MDL Number:
MFCD00025476 CAS Number: 88-97-1 EINECS: 201-871-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...