2-Propylcyclohexanone ≥98.0%

Supplier: TCI America

P1114-10ML
TCP1114-010MLEA 174.5 USD
TCP1114-010ML
2-Propylcyclohexanone ≥98.0%
2-Propylcyclohexanone

CAS Number: 94-65-5
MDL Number: MFCD00019464
Molecular Formula: C9H16O
Molecular Weight: 140.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 198
Specific Gravity (20/20): 0.91

Formula: C₉H₁₆O
MW: 140.23 g/mol
Boiling Pt: 198 °C
Density: 0.91 (20 °C)
MDL Number: MFCD00019464
CAS Number: 94-65-5
EINECS: 202-351-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR