2-(3-Phenylpropyl)pyridine ≥97.0%

Supplier: TCI America
Warning

P1327-5G
TCP1327-005GEA 117 USD
TCP1327-005G
2-(3-Phenylpropyl)pyridine ≥97.0%
2-(3-Phenylpropyl)pyridine

CAS Number: 2110-18-1
MDL Number: MFCD00191237
Molecular Formula: C14H15N
Molecular Weight: 197.28
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 171
Flash Point (°C): 100
Specific Gravity (20/20): 1.02

Formula: C₁₄H₁₅N
MW: 197.28 g/mol
Boiling Pt: 171 °C
Density: 1.02 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00191237
CAS Number: 2110-18-1
EINECS: 218-300-7
https://us.vwr.com/stibo/web/std.lang.all/19/85/28991985.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR