Phloxin B, BAKER ANALYZED®, Certified Stain for histology, J.T.Baker®

Synonyms: Sodium 2',4',5',7'-tetrabromo-4,5,6,7-tetrachloro-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)CyanosinePhloxineAcid Red 92Eosine Blue2′,4′,5′,7′-Tetrabromo-4,5,6,7-tetrachlorofluoresceinEosin S extra bluishCI 45410

Management of Change (MOC) category = NI

BAKRU029-03 BAKRU029-03
JTU029-3CS 683.32 USD
CAJTU029-3 JTU029-3
Phloxin B, BAKER ANALYZED®, Certified Stain for histology, J.T.Baker®
Phloxin B

Certified for Use in Histology (C.I. 45410)

Formula: C₂₀H₂Br₄Cl₄Na₂O₅
MW: 829.6344 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070627
CAS Number: 18472-87-2
EINECS: 242-355-6

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Total Dye Content, For Information Only
Absorbance Maximum, nm, For Information Only
Absorbance at Maximum (1.0 mg/200 mL in 50% C₂H₅OH and 0.01% Na₂CO₃, 1-cm path), For Information Only
Biological Test Passes Test
Certified by the Biological Stain Commission

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR