Periodic acid ≥99%, BAKER ANALYZED® ACS, J.T.Baker®

Danger

Synonyms: Orthoperiodic acidortho-Periodic acid

Management of Change (MOC) category = N

BAKRT146-03 BAKRT146-03 BAKRT146-05 BAKRT146-05
JTT146-5CS 1281.84 USD
CAJTT146-3 CAJTT146-5 JTT146-5 JTT146-3
Periodic acid ≥99%, BAKER ANALYZED® ACS, J.T.Baker®
Periodic acid

Lot analysis on label.

Formula: H₅IO₆
MW: 227.94 g/mol
Melting Pt: 122 °C
Density: 1.3875 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011344
CAS Number: 10450-60-9
EINECS: 233-937-0
UN: 3085
ADR: 5.1,I
Merck Index: 12,07312
https://us.vwr.com/stibo/web/std.lang.all/90/77/779077.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (H₅IO₆) 99.0 - 101.0 %
Insoluble Matter ≤0.01 %
Residue after Ignition ≤0.010 %
Sulfate (SO₄) ≤0.01 %
Other Halogens (as Cl) ≤0.01 %
Heavy Metals (as Pb) ≤0.005 %
Iron (Fe) ≤0.003 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR