Isoamyl acetate mixture of isomers ≥98%, J.T.Baker®

Danger

Synonyms: Isopentyl acetateAcetic acid isoamyl ester

Management of Change (MOC) category = N

BAKRT026-07
CAJTT026-7
Isoamyl acetate mixture of isomers ≥98%, J.T.Baker®
Isoamyl acetate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 142 °C (1013 hPa)
Melting Pt: –78 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008946
CAS Number: 123-92-2
EINECS: 204-662-3
UN: 1104
ADR: 3,III
Merck Index: 12,05125

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₇H₁₄O₂) (by GC)(total isomers) ≥98 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR