Thymol blue, BAKER ANALYZED® ACS, J.T.Baker®

Warning

Synonyms: ThymolsulphonphthaleinThymolsulfonphthalein3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

Management of Change (MOC) category = N

BAKRV856-02 V856-02 BAKRV856-01 V856-01
JTV856-1EA 250.2 USD
JTV856-1 JTV856-2 CAJTV856-1 CAJTV856-2
Thymol blue, BAKER ANALYZED® ACS, J.T.Baker®
Thymol blue

Lot analysis on label.

Formula: C₂₇H₃₀O₅S
MW: 466.6 g/mol
Melting Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00005869
CAS Number: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Clarity of Solution Passes Test
Visual Transition Intervals - pH…9.2 (Blue) Passes Test
Visual Transition Intervals - pH…1.2 (Pink) Passes Test
Visual Transition Intervals - pH…2.8 (Yellow) Passes Test
Visual Transition Intervals - pH…8.0 (Yellow) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR