2-Thiobarbituric acid ≥98%, BAKER ANALYZED®, J.T.Baker®

Synonyms: 4,6-Dihydroxy-2-mercaptopyrimidine

BAKRV774-05
CAJTV774-5
2-Thiobarbituric acid ≥98%, BAKER ANALYZED®, J.T.Baker®
2-Thiobarbituric acid
Formula: C₄H₄N₂O₂S
MW: 144.15 g/mol
Boiling Pt: 419 °C (1013 hPa)
Melting Pt: 235 °C
Density: 1.661 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006674
CAS Number: 504-17-6
EINECS: 207-985-8

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₄H₄N₂O₂S) ≥98 %
Residue after Ignition ≤0.10 %
Identification (by IR) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR