Tetramethylammonium chloride ≥98.0%, J.T. Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS
Danger

Synonyms: TMACl

V636-04 V636-04
JTV636-4CS 477.81 USD InStock
JTV636-4 CAJTV636-4
Tetramethylammonium chloride ≥98.0%, J.T. Baker®
Tetramethylammonium chloride
Formula: (CH₃)₄NCl
MW: 109.6 g/mol
Melting Pt: > 300 °C
Density: 1.169 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011628
CAS Number: 75-57-0
EINECS: 200-880-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₄H₁₂ClN) ≥ 98 %
Infrared Spectrum Passes Test
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR