1,1,2,2-Tetrachloroethane ≥98% (by GC), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US
Danger


Management of Change (MOC) category = N

BAKRV398-08 BAKRV398-08
JTV398-8EA 385.47 USD
CAJTV398-8 JTV398-8
1,1,2,2-Tetrachloroethane ≥98% (by GC), J.T.Baker®
1,1,2,2-Tetrachloroethane
Formula: Cl₂HCCHCl₂
MW: 167.85 g/mol
Boiling Pt: 146.3 °C (1013 hPa)
Melting Pt: –43 °C
Density: 1.595 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000848
CAS Number: 79-34-5
EINECS: 201-197-8
UN: 1702
ADR: 6.1,II
Merck Index: 12,09331
https://us.vwr.com/stibo/web/std.lang.all/53/34/725334.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CHCl₂CHCl₂) (by GC) ≥98 %
Appearance Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR