Trifluoroacetic acid ≥99.5% for HPLC, J.T.Baker®

Danger

Synonyms: TFA

BAKR9470-33 BAKR9470-07 BAKR9470-01 BAKR9470-00 BAKR9470-02 BAKR9470-07 BAKR9470-33 BAKR9470-01 9470-00 BAKR9470-02
JT9470-7CS 16162.56 USD
CAJT9470-7 CAJT9470-2 CAJT9470-1 JT9470-7 JT9470-33 JT9470-00 CAJT9470-33 JT9470-0 JT9470-2 JT9470-1
Trifluoroacetic acid ≥99.5% for HPLC, J.T.Baker®
Trifluoroacetic acid

Gradient tested for HPLC peptide and protein analysis. UV cutoff 210 nm.

Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 12,09812
https://us.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (F₃CCOOH) ≥99.5 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.15
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤210
Residue after Evaporation ≤10 ppm
Heavy Metals (as Pb) ≤10 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Gradient Elution Test (a.u.) - 215 nm ≤0.05

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR