Trichloroethylene ≥99% stabilized, CMOS for the electronics industry, J.T.Baker®

Danger

Synonyms: 1,1,2-TrichloroetheneTrichloroethene1,1,2-Trichloroethylene

BAKR9454-03
CAJT9454-3
Trichloroethylene ≥99% stabilized, CMOS for the electronics industry, J.T.Baker®
Trichloroethylene
Formula: ClCH=CCl₂
MW: 131.39 g/mol
Boiling Pt: 86.7 °C (1013 hPa)
Melting Pt: –73 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
EINECS: 201-167-4
UN: 1710
ADR: 6.1,III
Merck Index: 12,09769
https://us.vwr.com/stibo/web/std.lang.all/13/81/741381.jpg

Order Now

Specification Test Results

For Microelectronic Use
Assay (ClCH:CCl₂) (by GC) ≥ 99.0 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 5 ppm
Acidity (µeq/g) ≤ 0.1
Water (H₂O)(by coulometry) ≤ 0.005 %
Phosphate (PO₄) ≤ 0.10 ppm
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Arsenic and Antimony (as As) ≤ 10.0 ppb
Trace Impurities - Barium (Ba) ≤ 50.0 ppb
Trace Impurities - Boron (B) ≤ 10.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 20.0 ppb
Trace Impurities - Calcium (Ca) ≤ 20.0 ppb
Trace Impurities - Chromium (Cr) ≤ 20.0 ppb
Trace Impurities - Cobalt (Co) ≤ 20 ppb
Trace Impurities - Copper (Cu) ≤ 10.0 ppb
Trace Impurities - Gallium (Ga) ≤ 20.0 ppb
Trace Impurities - Germanium (Ge) ≤ 20 ppb
Trace Impurities - Gold (Au) ≤ 20 ppb
Heavy Metals (as Pb) ≤ 100 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Lead (Pb) ≤ 10.0 ppb
Trace Impurities - Lithium (Li) ≤ 50.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 30.0 ppb
Trace Impurities - Manganese (Mn) ≤ 20.0 ppb
Trace Impurities - Nickel (Ni) ≤ 10.0 ppb
Trace Impurities - Potassium (K) ≤ 100 ppb
Trace Impurities - Silicon (Si) ≤ 50 ppb
Trace Impurities - Silver (Ag) ≤ 20.0 ppb
Trace Impurities - Sodium (Na) ≤ 100.0 ppb
Trace Impurities - Strontium (Sr) ≤ 20.0 ppb
Trace Impurities - Tin (Sn) ≤ 20.0 ppb
Trace Impurities - Titanium (Ti) ≤ 10.0 ppb
Trace Impurities - Zinc (Zn) ≤ 20.0 ppb
Particle Count - 1.0 µm and greater ≤ 10 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR