Tetrahydrofuran, ultra-low water content ≥99.8%, BakerDRY™, J.T.Baker®

Danger

Synonyms: OxolaneTHFCyclotetramethylene oxideTetramethylene oxide1,4-epoxy-ButaneButane alpha,delta-oxideOxacyclopentane

Management of Change (MOC) category = NI

BAKR9447-10 BAKR9447-12 BAKR9447-10 BAKR9447-12
JT9447-10CS 5639.06 USD
JT9447-10 JT9447-12 CAJT9447-10 CAJT9447-12
Tetrahydrofuran, ultra-low water content ≥99.8%, BakerDRY™, J.T.Baker®
Tetrahydrofuran

Clear, colorless liquid; ether-like odor. For use in organic synthesis, including organometallic synthesis. Contains BHT as a preservative.

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://us.vwr.com/stibo/web/std.lang.all/95/19/789519.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (by GC, corrected for water) ≥99.8 %
ACS - Color (APHA) ≤10
ACS - Peroxide (as H₂O₂) (at time of manufacture) ≤0.015 %
ACS - Residue after Evaporation ≤0.03 %
ACS - Water (by KF, coulometric) ≤10.0 ppm
Stabilizer (BHT) 100 - 300 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR