Tetrahydrofuran, ultra-low water content ≥99.8% (by GC, corrected for water content), BakerDRY™ for organic synthesis, J.T.Baker®

Danger

Synonyms: OxolaneTHFCyclotetramethylene oxideTetramethylene oxide1,4-epoxy-ButaneButane alpha,delta-oxideOxacyclopentane

Management of Change (MOC) category = NI

BAKR9446-99 BAKR9446-7N BAKR9446-5A BAKR9446-5N BAKR9446-YY BAKR9446-10 BAKR9446-7Y BAKR9446-50 BAKR9446-X9 BAKR9446-YN BAKR9446-7N BAKR9446-1M BAKR9446-1M BAKR9446-4R BAKR9446-U3 BAKR9446-X7 BAKR9446-YN BAKR9446-99 BAKR9446-12 BAKR9446-9B BAKR9446-98 BAKR9446-10 BAKR9446-ZN BAKR9446-50 BAKR9446-8W BAKR9446-8W BAKR9446-X9 BAKR9446-5A BAKR9446-YY BAKR9446-7Y BAKR9446-12 BAKR9446-U3 BAKR9446-4R BAKR9446-9B BAKR9446-ZN BAKR9446-X7 BAKR9446-98 BAKR9446-5N
JT9446-1MCS 3842.31 USD
JT9446-1M CAJT9446-50 JT9446-U3 CAJT9446-4R CAJT9446-X7 CAJT9446-10 CAJT9446-8W CAJT9446-98 CAJT9446-99 CAJT9446-YY JT9446-7Y CAJT9446-12 CAJT9446-X9 CAJT9446-9B CAJT9446-5A JT9446-YN CAJT9446-ZN CAJT9446-7N CAJT9446-5N JT9446-50 CAJT9446-1M JT9446-4R JT9446-10 JT9446-98 JT9446-8W JT9446-X7 JT9446-99 JT9446-YY CAJT9446-U3 JT9446-X9 JT9446-12 CAJT9446-7Y JT9446-9B JT9446-5A JT9446-ZN JT9446-7N CAJT9446-YN JT9446-5N
Tetrahydrofuran, ultra-low water content ≥99.8% (by GC, corrected for water content), BakerDRY™ for organic synthesis, J.T.Baker®
Tetrahydrofuran

Specifically manufactured for use in organic, organometallic, and oligonucleotide synthesis requiring low water solvents.

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://us.vwr.com/stibo/web/std.lang.all/95/19/789519.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (by GC, corrected for water) ≥99.8 %
ACS - Color (APHA) ≤10
ACS - Peroxide (as H₂O₂) (at time of manufacture) ≤0.015 %
ACS - Residue after Evaporation ≤1 ppm
ACS - Water (by KF, coulometric) ≤10.0 ppm
Packaged under Argon

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR