Thioacetamide ≥99.0%, crystals, BAKER ANALYZED®, J.T.Baker®

Danger

Synonyms: Ethanethioamide

Management of Change (MOC) category = N

BAKR8984-04 8984-01 BAKR8984-04
JT8984-4CS 1473.81 USD
CAJT8984-1 JT8984-4 CAJT8984-4
Thioacetamide ≥99.0%, crystals, BAKER ANALYZED®, J.T.Baker®
Thioacetamide

Crystals. Lot analysis on label.

Formula: CH₃CSNH₂
MW: 75.13 g/mol
Melting Pt: 113…114 °C
Density: 1.336 g/cm³ (20 °C)
MDL Number: MFCD00008070
CAS Number: 62-55-5
EINECS: 200-541-4
Merck Index: 12,09453
https://us.vwr.com/stibo/web/std.lang.all/72/66/2297266.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃CSNH₂) ≥99.0 %
Melting Point 111.0 - 114.0 °C
Residue after Ignition ≤0.050 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR