Petroleum spirit, 30…60 °C, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Danger

Synonyms: Petroleum etherGasolinePetrol

Management of Change (MOC) category = NI

BAKR9265-H1 BAKR9265-03 BAKR9265-H1 BAKR9265-03
JT9265-H1CS 62073.97 USD
CAJT9265-H1 JT9265-H1 JT9265-3 CAJT9265-3
Petroleum spirit, 30…60 °C, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Petroleum spirit
Density: ~0.68 kg (20 °C)
Flash Pt: −18 °C
MDL Number: MFCD00081849
CAS Number: 8032-32-4
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL) ≤5
Color (APHA) ≤10
Residue after Evaporation ≤5 ppm
Water (by KF, coulometric) ≤0.05 %
Boiling Range(Lower) 30 - 60 °C
Boiling Range(Upper) 30 - 60 °C
ECD-Sensitive Impurities (as Ethylene Dibromide)
MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR