Silica gel, Pore size: 30 Å ≥99.7% (dry basis), SilicAR (100-200 Mesh)(75-150 Microns), Macron Fine Chemicals™


Management of Change (MOC) category = N

MACRV152-02 MACRV152-02 MACRV152-13 V152-13
MKV15213CS 14773.74 USD
MKV15213 CAMKV15213 MKV152-02 JTV152-02
Silica gel, Pore size: 30 Å ≥99.7% (dry basis), SilicAR (100-200 Mesh)(75-150 Microns), Macron Fine Chemicals™
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (SiO₂, dry basis) ≥99.7 %
pH of 5% Slurry in Water 5.5 - 7.0
Total Volatiles ≤10.0 %
Surface Area (m²/g) 430 - 530
On U.S. No. 60 Sieve
On U.S. No. 80 Sieve ≤1.2 %
On U.S. No. 100 Sieve ≤5.0 %
Thru U.S. No. 200 Sieve ≤15.0 %
Appearance (fine, white powder)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR