Strontium nitrate, anhydrous ≥99% ACS, Macron Fine Chemicals™

Warning

MACR8256-04
CAMK825604
Strontium nitrate, anhydrous ≥99% ACS, Macron Fine Chemicals™
Strontium nitrate

Granular.

Formula: Sr(NO₃)₂
MW: 211.63 g/mol
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 570 °C
Density: 2.113 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1,III
Merck Index: 12,09007

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Sr(NO₃)₂) ≥ 99.0 %
ACS - Barium (Ba) ≤ 0.05 %
ACS - Calcium (Ca) ≤ 0.05 %
ACS - Chloride (Cl) ≤ 0.002 %
ACS - Heavy Metals (as Pb) ≤ 5 ppm
ACS - Insoluble Matter ≤ 0.01 %
ACS - Iron (Fe) ≤ 5 ppm
ACS - Loss on Drying at 105°C ≤ 0.1 %
ACS - Magnesium (Mg) ≤ 0.10 %
ACS - Sodium (Na) ≤ 0.10 %
ACS - Sulfate (SO₄) ≤ 0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 7.0
Appearance (crystalline)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR