Sodium peroxide ≥93.0%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: di-Sodium peroxide

Management of Change (MOC) category = N

MACR7864-04 MACR7864-04
MK786404CS 4096.06 USD
CAMK786404 MK786404
Sodium peroxide ≥93.0%, granular, AR® ACS, Macron Fine Chemicals™
Sodium peroxide

Granular.

Formula: Na₂O₂
MW: 77.98 g/mol
Melting Pt: 660 °C
Density: 2.805 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I
Merck Index: 12,08800

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Na₂O₂) (by KMnO₄ titrn) ≥93.0 %
ACS - Chloride (Cl) ≤0.002 %
ACS - Heavy Metals (as Pb) ≤0.002 %
ACS - Iron (Fe) ≤0.005 %
ACS - Phosphate (PO₄) ≤0.0005 %
ACS - Sulfate (SO₄) ≤0.001 %
Appearance (white to pale yellow, hygroscopic granules)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR