Sodium hydrogen carbonate 99.7-100.3% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Sodium bicarbonate
Management of Change (MOC) category = N
MACR7412-20
MACR7412-06
MACR7412-09
7412-12
MACR7412-20
MACR7412-12
MACR7412-28
MACR7412-28
MACR7412-09
MK741212CS
304.19
USD
MK741212
CAMK7412-28
MK741220
CAMK7412-09
JT7412-09
CAMK741220
MK7412-28
MK741206
CAMK741212
Sodium hydrogen carbonate 99.7-100.3% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Sodium hydrogen carbonate
Formula:
NaHCO₃ MW: 84.01 g/mol Melting Pt: 270 °C Density: 2.159 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003528 CAS Number: 144-55-8 EINECS: 205-633-8 Merck Index: 12,08726 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (NaHCO₃) (by acidimetry) | 99.7 - 100.3 % |
ACS - Ammonium (NH₄) | ≤5 ppm |
ACS - Calcium (Ca) | ≤0.02 % |
ACS - Chloride (Cl) | ≤0.003 % |
ACS - Heavy Metals (as Pb) | ≤5 ppm |
ACS - Insoluble Matter | ≤0.015 % |
ACS - Iron (Fe) | ≤0.001 % |
ACS - Magnesium (Mg) | ≤0.005 % |
ACS - Phosphate (PO₄) | ≤0.001 % |
ACS - Potassium (K) | ≤0.005 % |
ACS - Sulfur Compounds (as SO₄) | ≤0.003 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...