Succinic acid ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™

Danger

Synonyms: 1,4-Butanedioic acid

Management of Change (MOC) category = NI

MACR2860-06 2860-06 MACR2860-04
MK286006CS 4369.71 USD
CAMK286006 MK286006 MK286004
Succinic acid ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Succinic acid
Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 182 °C
Density: 1.56 g/cm³ (20 °C)
Flash Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8,III
Merck Index: 12,09038
https://us.vwr.com/stibo/web/std.lang.all/01/38/2300138.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HOCOCH₂CH₂COOH) ≥99.0 %
ACS - Appearance (white granules) Passes Test
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤5 ppm
ACS - Melting Point 185.0 - 191.0 °C
ACS - Nitrogen Compounds (as N) ≤0.001 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Residue after Ignition ≤0.02 %
ACS - Solubility (10 in 150) Passes Test
ACS - Sulfate (SO₄) ≤0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR