Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Jumbo Temperature/Humidity Meter

Supplier: VWR International
VWR®
4184 VC3007 VC4184
Unit features an external thermometer probe with 3.1m (10') cable and a 2.9 cm (11/8") high digital readout that continuously and simultaneously displays ambient humidity and ambient and external probe temperatures.

  • Traceable®
  • Dual recall memory
  • Reads °F and °C

Display is updated once a second and is readable from up to 10.7m (35'). Fast-response, solid-state sensors show condition changes in less than 30 seconds. The minimum and maximum memories may be displayed or cleared at any time with the touch of a button. External temperature sensor performs accurately under water. Self-contained unit requires no adjustment or maintenance. Case is chemical and shock-resistant ABS plastic.

Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. Unit meets federal requirements for CLIA specification 493.1207 for monitoring temperature and humidity.

Ordering information: One-year warranty. Supplied with probe and cable, flip-open stand, wall mount, and AAA alkaline battery.



 

ORDER

Expand All / Collapse All

Meters
L x H x D VWR Catalog Number Unit Price Quantity
9.8 x 10.8 x 1.9 cm (327/32 x 41/4 x 3/4") 61161-378 Each Retrieving

VWRCATALYST Re-Calibration Service (Service Only, Without Hygrometer)
Description Includes VWR Catalog Number Unit Price Quantity
Original Manufacturer Calibration Per Hygrometer ISO 17025 NIST Certification, Original Manufacturer Re-Calibration & no extra shipping fee 10754-850 Each Retrieving
Two-Point Wide Range Calibration Per Hygrometer ISO 17025 NIST Certification, Two Point Calibration for Humidity and one Point for Temperature Per Manufacturer and no extra shipping fee 10754-734 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Temperature Accuracy ±1°C
Temperature Resolution
Humidity Range 25–95% RH
Humidity Resolution 1% RH
Humidity Accuracy ±2–4%
Temperature Range, External Probe –50 to 70°C (–58 to 158°F)
Temperature Range, Ambient Sensor –5 to 50°C (23 to 122°F)

SERVICES


Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital hygrometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. You will receive a protective box to ship your instrument to VWR's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request. Hygrometer not included.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty