Amidosulfonic acid ≥99%, crystals, Macron Fine Chemicals™

Danger

Synonyms: Sulfamic acidAminosulfonic acid Sulfamidic acid Amidosulfonic acid

Management of Change (MOC) category = N

1931-59 MACR1931-05 MACR1931-59
MK1931-05EA 363.2 USD
MK1931-05 CAMK193159 MK193159
Amidosulfonic acid ≥99%, crystals, Macron Fine Chemicals™
Amidosulphonic acid
Formula: H₃NO₃S
MW: 97.09 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 205 °C
Density: 2.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8,III
Merck Index: 12,09090
https://us.vwr.com/stibo/web/std.lang.all/75/87/2297587.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Solubility (5 in 100) Passes Test
Identification Passes Test
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.0005 %
Appearance (colorless or white crystals)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR