Amidosulfonic acid 99.5-100.3%, BAKER ANALYZED® ACS, J.T.Baker®

Danger

Synonyms: Aminosulphonic acidAmidosulphonic acidSulphamic acidAmidosulfuric AcidAmidosulphuric AcidSulphamidic acid

Management of Change (MOC) category = N

BAKRV145-05 BAKRV145-07
JTV145-5EA 121.09 USD
JTV145-5 JTV145-7
Amidosulfonic acid 99.5-100.3%, BAKER ANALYZED® ACS, J.T.Baker®
Amidosulphonic acid

Lot analysis on label.

Formula: H₃NO₃S
MW: 97.09 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 205 °C
Density: 2.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8,III
Merck Index: 12,09090
https://us.vwr.com/stibo/web/std.lang.all/75/87/2297587.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (NH₂SO₃H) (dried basis) 99.3 - 100.3 %
Insoluble Matter ≤0.01 %
Residue after Ignition ≤0.010 %
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.05 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR