Schiff reagent (Hotchkiss & McManus), J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS US
|
Danger
|
Management of Change (MOC) category = NI
BAKRU973-01
BAKRU973-01
JTU973-1CS
2514.71
USD
JTU973-1
CAJTU973-1
Schiff reagent (Hotchkiss & McManus), J.T.Baker®
Schiff reagent
For Periodic Acid-Schiff Reaction.
Contains 4.54 g/L basic fuchsin, 4.543 g/L sodium bisulphite, and 7.5 ml/L hydrochloric acid in water.
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
pH at 25°C | 1.1 - 1.5 |
Absorbance at 545 nm (10-mm path vs water) | ≤0.020 |
Sensitivity for Aldehyde | Passes Test |
Suitability for Periodic Acid-Schiff Staining | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...