Sodium sulfite ≥95%, purified, exsiccated, J.T.Baker®


Management of Change (MOC) category = N

BAKR3888-01 BAKR3888-01
JT3888-1CS 335.13 USD
CAJT3888-1 JT3888-1
Sodium sulfite ≥95%, purified, exsiccated, J.T.Baker®
Sodium sulphite
Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: > 500 °C
Density: 2.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 12,08831

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (Na₂SO₃) ≥95 %
Description Passes Test
Identification Passes Test
Solubility in H₂O Passes Test
Thiosulfate (S₂O₃) Passes Test
Heavy Metals (as Pb) ≤0.002 %
Trace Impurities - Arsenic (As) ≤8.000 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR