Sodium silicate ∼41° Baume in aqueous solution, J.T.Baker®

Danger


Management of Change (MOC) category = N

3877-01 BAKR3877-01
JT3877-1CS 1202.15 USD
CAJT3877-1 JT3877-1
Sodium silicate ∼41° Baume in aqueous solution, J.T.Baker®
Sodium silicate
Formula: Na₂O(SiO₂)x·xH₂O
MDL Number: MFCD00003492
CAS Number: 1344-09-8
EINECS: 215-687-4
UN: 3266
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Specific Gravity at 25°/25°C 1.35 - 1.42
Viscosity approximately 180 cps.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR