Sodium fluoride 98.0-102.0% (dry basis), powder USP, J.T.Baker®

Danger


Management of Change (MOC) category = HR

BAKR3689-08 BAKR3689-09 BAKR3689-08 BAKR3689-QM BAKR3689-05 BAKR3689-01 BAKR3689-09
JT3689-8EA 1915.84 USD
CAJT3689-5 CAJT3689-8 CAJT3689-09 JT3689-8 JT3689-QM JT3689-09 JT3689-1
Sodium fluoride 98.0-102.0% (dry basis), powder USP, J.T.Baker®
Sodium fluoride
Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1700 °C (1013 hPa)
Melting Pt: 993 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Merck Index: 12,08762
https://us.vwr.com/stibo/web/std.lang.all/06/30/16640630.jpg

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acidity or Alkalinity Passes Test
USP - Loss on Drying ≤1.0 %
USP - Fluosilicate (volume of 0.10N NaOH required) ≤1.5 ml
USP - Chloride (Cl) ≤0.012 %
USP - Assay (NaF) (dried basis) 98.0 - 102.0 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR