Salicylic acid ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: o-Hydroxybenzoic acid2-Hydroxybenzoic acid

Management of Change (MOC) category = N

BAKR0300-05 BAKR0300-01 0300-05 BAKR0300-01
JT0300-1CS 726.66 USD
JT0300-1 JT0300-5 CAJT0300-1 CAJT0300-5
Salicylic acid ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Salicylic acid

  • Lot analysis on label

Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 158…161 °C
Density: 1.443 g/cm³ (20 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 12,08484
https://us.vwr.com/stibo/web/std.lang.all/99/00/749900.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₇H₆O₃) ≥99.0 %
Melting Point 158.0 - 161.0 °C
Residue after Ignition ≤0.010 %
Substances Darkened by H₂SO₄ Passes Test
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.002 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR