Phenol ≥99% USP, fused, J.T.Baker®
|
Danger
|
Synonyms:
Carbolic acid
, Hydroxybenzene
, Phenylalcohol
Management of Change (MOC) category = R
BAKR2862-05
BAKR2862-05
JT2862-5CS
2048.27
USD
JT2862-5
CAJT2862-5
Phenol ≥99% USP, fused, J.T.Baker®
Phenol
Fused. Meets USP specifications.
Formula:
C₆H₅OH MW: 94.11 g/mol Boiling Pt: 181.7 °C (1013 hPa) Melting Pt: 40.8 °C Density: 1.0722 g/cm³ (20 °C) Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002143 CAS Number: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1,II REACH: 01-2119471329-32 Merck Index: 12,07390 |
Specification Test Results
Meets U.S.P Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Congealing Temperature | ≥39 °C |
USP - Clarity of Solution and Reaction | Passes Test |
USP - Water (H₂O) | ≤0.5 % |
USP - Nonvolatile Residue | ≤0.05 % |
USP - Assay (C₆H₆O) (anhydrous basis) | 99.0 - 100.5 % |
USP - Benzene | ≤2.0 ppm |
Product contains about 20 ppm Citric Acid as a preservative. | |
No Class 2 or 3 solvents are expected to be present. The Class 1 | |
solvent Benzene is used as a raw material in a precursor to phenol. The product is tested for any presence of benzene and the result | |
reported on the Certificate of Analysis. | |
Elemental Impurities (USP 232) - Information on elemental impurities for | |
this product is available on the associated Product Regulatory Data | |
Sheet and elemental impurity profile report. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...