Sodium hydrogen carbonate 99.7-100.3%, GR ACS, Supelco®
Synonyms:
Sodium bicarbonate
SX0320-3
EM-SX0320-3CS
375.48
USD
EM-SX0320-3
Sodium hydrogen carbonate 99.7-100.3%, GR ACS, Supelco®
Sodium hydrogen carbonate
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Formula:
NaHCO₃ MW: 84.01 g/mol Melting Pt: 270 °C Density: 2.159 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003528 CAS Number: 144-55-8 EINECS: 205-633-8 Merck Index: 12,08726 |
Specification Test Results
Assay (dried basis) | 99.7-100.3% |
Ammonium (NH4) | 5ppm max |
Calcium (Ca) | 0.02% max |
Chloride (Cl) | 0.003% max |
Heavy metals (as Pb) | 5ppm max |
Insoluble matter | 0.015% max |
Iron (Fe) | 0.001% max |
Magnesium (Mg) | 0.005% max |
Phosphate (PO4) | 0.001% max |
Potassium (K) | 0.005% max |
Sulfur compounds (as SO4) | 0.003% max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...