Silver nitrate, crystals, GR ACS, Supelco®

Danger

Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

SX0205-7 SX0205-5 SX0205-7 SX0205-3 SX0205-3 SX0205-5
EM-SX0205-5EA 2243.06 USD
EM-SX0205-5 EM-SX0205-7 CASX0205-7 CASX0205-5 CASX0205-3 EM-SX0205-3
Silver nitrate, crystals, GR ACS, Supelco®
Silver nitrate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 210 °C
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://us.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg

Order Now

Specification Test Results

Assay 99.0% min
Chloride (Cl) 5ppm max
Clarity of solution To pass test
Color Colorless
Copper (Cu) 2ppm max
Form Crystals
Free acid To pass test
Iron (Fe) 2ppm max
Lead (Pb) 0.001% max
Substances not precipitated by HCl 0.01% max
Sulfate (SO4) 0.002% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR