Unit uses laser targeting for precise temperature measurement.
- Traceable®
- 8:1 field of view
- Backlit display
- Can freeze display reading to hold data
- Memory functions retains past maximum/minimum readings
Since the infrared output of some objects is unique, keys permit adjusting the emissivity from 0.3 to 1.0
Features include backlighting, data hold (freezes the display reading), laser, memory for maximum/minimum/average readings, nine memories, high/low audible alarm, differential readings, low-battery indicator, and a °C/°F button.
Field of view is 8:1, indicating that the thermometer can read a spot the size of 5 cm (2") from 40.6 cm (16") away.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with wrist strap, soft-sided carrying case, and 9V battery.