Reactive blue 4, OR, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

MACR8132-03
MK813203EA 1300.08 USD
MK813203
Reactive blue 4, OR, Macron Fine Chemicals™
Reactive blue 4
Formula: C₂₃H₁₄Cl₂N₆O₈S₂
MW: 637.44 g/mol
MDL Number: MFCD00001218
CAS Number: 13324-20-4

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Identification Passes Test
UV Absorbance at 280 nm and 253 nm Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR