Rosolic acid (Aurin), powder biological stain C.I. 43800, Sigma-Aldrich®

Synonyms: 4-(Bis(4-hydroxyphenyl)methylene)cyclohexa-2,5-dien-1-one

RX0175-1
CARX0175-1
Rosolic acid (Aurin), powder biological stain C.I. 43800, Sigma-Aldrich®
Rosolic acid (Aurin)
Formula: C₁₉H₁₄O₃
MW: 290.32 g/mol
MDL Number: MFCD00001624
CAS Number: 603-45-2
EINECS: 210-041-8

Order Now

Specification Test Results

Color Red
Extinction coefficient 45000 min
Form Powder
Loss on drying 5.0% max
Wavelength of max. absorbance 480-485nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR