1-Octanol ≥99.5%
Supplier: TCI America
|
Warning
|
Synonyms:
n-Octanol
O0212-500ML
TCO0212-500MLEA
277.99
USD
TCO0212-500ML
1-Octanol ≥99.5%
1-Octanol
[for Determining Partition Coefficients]
CAS Number: 111-87-5
MDL Number: MFCD00002988
Molecular Formula: C8H18O
Molecular Weight: 130.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 194
Melting point (°C): -16
Flash Point (°C): 83
Specific Gravity (20/20): 0.83
Other Product Names / Synonyms: n-Octyl Alcohol
Formula:
H₃C(CH₂)₆CH₂OH MW: 130.23 g/mol Boiling Pt: 194 °C Melting Pt: –16 °C Density: 0.83 (20 °C) Flash Pt: 83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 EINECS: 203-917-6 Merck Index: 12,06849 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...