Nitric acid fuming ≥90.0%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIALS US
Danger


Management of Change (MOC) category = N

MACR2713-41 MACR2713-41
MK271341EA 850.24 USD
CAMK271341 MK271341
Nitric acid fuming ≥90.0%, AR® ACS, Macron Fine Chemicals™
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 87 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,I
REACH: 01-2119487297-23
Merck Index: 12,06672

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HNO₃) (by acidimetry) ≥90.0 %
Dilution Test Passes Test
ACS - Residue after Ignition ≤0.002 %
ACS - Dissolved Oxides (as N₂O₃) ≤0.1 %
ACS - Chloride (Cl) ≤0.7 ppm
ACS - Sulfate (SO₄) ≤5 ppm
ACS - Arsenic (As) ≤0.3 ppm
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤2 ppm
Appearance (colorless to slightly yellow-brown solution)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR