Nicotinic acid (Niacin), J.T.Baker®

Warning

Synonyms: Pyridine-3-carboxylic acid3-Pyridinecarboxylic acidVitamin B33-Picolinic acidNicotinic acidNiacin

Management of Change (MOC) category = N

BAKRR763-07
CAJTR763-7
Nicotinic acid (Niacin), J.T.Baker®
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 142 °C
Density: 1.473 g/cm³ (20 °C)
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 12,06612

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Melting Point 236 - 238 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR