N-(1-Naphthyl)ethylenediamine dihydrochloride, BAKER ANALYZED® ACS, J.T.Baker®

Warning

Synonyms: N-1-NaphthylethylenediamineN-2-Aminoethyl-1-naphthylamine dihydrochlorideMarshall's Reagent

Management of Change (MOC) category = N

BAKRR701-03 BAKRR701-05 R701-05 BAKRR701-03
JTR701-5CS 2655.42 USD
CAJTR701-5 CAJTR701-3 JTR701-5 JTR701-3
N-(1-Naphthyl)ethylenediamine dihydrochloride, BAKER ANALYZED® ACS, J.T.Baker®
N-2-Aminoethyl-1-naphthylamine dihydrochloride

For nitrogen dioxide determination (ASTM D-1607). Lot analysis on label.

Formula: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS Number: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://us.vwr.com/stibo/web/std.lang.all/64/71/2346471.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Sensitivity for Sulfanilamide Passes Test
Solubility Passes Test
ACS - Water (H₂O)(by Karl Fischer titrn) ≤5 %
ACS - Suitability for Nitrogen Dioxide Determination (ASTM D-1607) Passes Test
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR