Methyl butyrate ≥99.0%

Supplier: TCI America

Synonyms: Butanoic acid methyl ester Butyric acid methyl ester

B0763-500ML B0763-25ML
TCB0763-500MLEA 51.77 USD
TCB0763-500ML TCB0763-25ML
Methyl butyrate ≥99.0%
Methyl butyrate

CAS Number: 623-42-7
MDL Number: MFCD00009391
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Flash Point (°C): 13
Specific Gravity (20/20): 0.90


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Butyric Acid Methyl Ester

Formula: C₅H₁₀O₂
Boiling Pt: 101 °C
Melting Pt: –84 °C
Density: 0.90 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00009391
CAS Number: 623-42-7
EINECS: 210-792-1
UN: 1237
ADR: 3,II
Merck Index: 12,06114
https://us.vwr.com/stibo/web/std.lang.all/26/89/28932689.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR