Trifluoroacetic acid 0.05% (v/v) in water mobile phase for HPLC, Supelco®
Synonyms:
TFA
TX1282P-1
CA-TX1282P-1
Trifluoroacetic acid 0.05% (v/v) in water mobile phase for HPLC, Supelco®
Trifluoroacetic acid
HPLC
Formula:
C₂HF₃O₂ MW: 114.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 EINECS: 200-929-3 |
Specification Test Results
Assay (trifluoroacetic acid) | 0.045-0.055% (v/v) |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Form | Clear liquid |
Gradient at 210 nm | 0.03AU max |
Gradient at 280 nm | 0.01AU max |
Residue after evaporation | 5ppm max |
UV Abs. at 205 nm | 1.00AU max |
UV Abs. at 210 nm | 0.70AU max |
UV Abs. at 230 nm | 0.30AU max |
UV Abs. at 250 nm and higher | 0.05AU max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...