Mercury(II) chloride 5% (w/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Mercury dichlorideMercuric chlorideMercury Chloride

R4650000-500A
RC4650-16EA 126.7 USD
RC4650-16
Mercury(II) chloride 5% (w/v) in aqueous solution
Mercury (II) chloride

Clear, colorless, odorless liquid.

Formula: HgCl₂
MW: 271.5 g/mol
MDL Number: MFCD00011041
CAS Number: 7487-94-7
UN: 1624
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR