Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3%, BAKER ANALYZED® ACS, J.T.Baker®

Danger

Synonyms: Ethylene glycol monomethyl ether

Management of Change (MOC) category = N

BAKRP784-08 BAKRP784-08 BAKRP784-07 BAKRP784-07
JTP784-7CS 1500.1 USD
JTP784-7 CAJTP784-7 CAJTP784-8 JTP784-8
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3%, BAKER ANALYZED® ACS, J.T.Baker®
Ethylene glycol monomethyl ether (2-Methoxyethanol)

Lot analysis on label.

Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 124.6 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.964 g/cm³ (20 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₃H₈O₂) (by GC, corrected for water) ≥99.3 %
Color (APHA) ≤10
Titrable Acid (meq/g) ≤0.002
Water (H₂O)(by Karl Fischer titrn) ≤0.1 %
pH (80% in Water), For Information Only
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR