Malachite green oxalate, BAKER ANALYZED®, Certified Stain for histology, for bacteriology, J.T.Baker®

Danger

Synonyms: Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalateN-[4-[[4-(Dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-methanaminium, ethanedioate, ethanedioate(2:2:1)Basic Green 4N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate

BAKRP450-03
CAJTP450-3
Malachite green oxalate, BAKER ANALYZED®, Certified Stain for histology, for bacteriology, J.T.Baker®
Malachite green oxalate

Certified for Use in Histology and Bacteriology (Staining) (C.I. 42000)

Formula: C₅₂H₅₄N₄O₁₂
MW: 927.02 g/mol
Melting Pt: ∼159 °C
Storage Temperature: Ambient
MDL Number: MFCD00011766
CAS Number: 2437-29-8
EINECS: 219-441-7
UN: 2928
ADR: 6.1 (8),II
Merck Index: 13,05722
https://us.vwr.com/stibo/web/std.lang.all/75/29/2297529.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Total Dye Content, For Information Only
Absorbance Maximum, nm, For Information Only
Absorbance at Maximum (0.6 mg/200 mL in H₂O, 1-cm path), For Information Only
Biological Test Passes Test
Certified by the Biological Stain Commission

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR