Methyl orange 90-100%, powder, BAKER ANALYZED® ACS C.I.13025, J.T.Baker®

Danger

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium saltMethyl orange sodium saltHelianthine4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt

Management of Change (MOC) category = N

BAKR2694-04 2694-04
JT2694-4CS 882.91 USD
JT2694-4 CAJT2694-4
Methyl orange 90-100%, powder, BAKER ANALYZED® ACS C.I.13025, J.T.Baker®
Methyl orange

Powder. Lot analysis on label.

Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Boiling Pt: ∼90 °C (1013 hPa)
Melting Pt: > 300 °C
Density: ∼1 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
UN: 2811
ADR: 6.1,III
Merck Index: 12,06180

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Clarity of Solution Passes Test
Visual Transition Interval - pH…3.2 (Red) Passes Test
Visual Transition Interval - pH…4.4 (Yellow) Passes Test
Appearance (white to pale yellow crystals or powder)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR