Paraffin Wax Dispenser, Electrothermal
MH8523BX1 MH8523BX1
15156-100EA 4545.76 USD
15156-100 CA15156-100
Paraffin Wax Dispenser, Electrothermal
Immediate “Wax on Tap” for pathology/histology laboratories.
Thermally insulated tap heater with independent temperature control completely eliminates problem of wax solidifying at the tap. Aluminum housing with stove enameled black and white anodized interior, black metal lid, and propylene base. Thermostat and tap heater built-in. Tap is swivel lever nondrip with replaceable filter. Has power on, bath on, tap heater on, and over-temperature indicators. Equipped with safety over-temperature cutout at 105°C (221°F) and nonresettable thermal fuse. 3-core cable power connection.
Ordering information: For 230V units, contact your VWR sales representative.