Manganese(II) carbonate Mn ≥ 43.0%, powder, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®


Management of Change (MOC) category = N

BAKR2536-R BAKR2536-07 2536-07
JT2536-REA 8281.82 USD
JT2536-R CAJT2536-7 JT2536-7
Manganese(II) carbonate Mn ≥ 43.0%, powder, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Manganese (II) carbonate

Powder. Lot analysis on label.

Formula: CMnO₃
MW: 114.95 g/mol
Density: 3.12 g/cm³ (20 °C)
MDL Number: MFCD00011116
CAS Number: 598-62-9
EINECS: 209-942-9
Merck Index: 12,05766

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (as Mn) ≥43.0 %
Insoluble in HCl ≤0.01 %
Calcium (Ca) ≤0.01 %
Chloride (Cl) ≤0.02 %
Magnesium (Mg) ≤0.01 %
Potassium (K) ≤0.01 %
Sulfate (SO₄) ≤0.005 %
Other Heavy Metals (as Pb) ≤0.005 %
Iron (Fe) ≤0.02 %
Sodium (Na) ≤0.02 %
Zinc (Zn) ≤0.05 %
Average Particle Diameter, µm (APD) ≤3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR