Mercury(II) acetate ≥99.0%, GR ACS, Supelco®

Danger

Synonyms: Acetic acid mercury (II) saltMercury diacetateMercuric acetate

MX0335-2 MX0335-2
EM-MX0335-2CS 2023 USD
CAMX0335-2 EM-MX0335-2
Mercury(II) acetate ≥99.0%, GR ACS, Supelco®
Mercury (II) acetate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: (H₃CCOO)₂Hg
MW: 318.68 g/mol
Melting Pt: 178…180 °C
Density: 3.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898

Order Now

Specification Test Results

Assay (complexometric) 99.0% min
Chloride (Cl) 0.005% max
Other heavy metals 0.002% max
Insoluble matter 0.01% max
Iron (Fe) 0.001% max
Mercury (I) (as Hg) 0.3% max
Nitrate (NO3) 0.005% max
Residue after reduction 0.02% max
Sulfate (SO4) 0.005% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR