γ-(N-Morpholino)propanesulfonic acid (MOPS), OmniPur®, Millipore®

Synonyms: 3-(N-Morpholino)propanesulphonic acid3-Morpholinopropane-1-sulfonic acid3-Morpholinopropane-1-sulphonic acid3-(N-Morpholino)propanesulfonic acid

6310-100GM 6320-1KG 6340-25KG 6310-100GM
EM-6310EA 165.44 USD
CA-EM6310 EM-6310 CA6320 EM-6340
γ-(N-Morpholino)propanesulfonic acid (MOPS), OmniPur®, Millipore®
γ-(N-Morpholino)propanesulphonic acid (MOPS)

DNase-, RNase-, protease-, and phosphatase-free.

Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Melting Pt: > 250 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 12,06346

Order Now

Specification Test Results

A (260 nm, 0.1 M, water) 0.05AU max
A (280 nm, 0.1 M, water) 0.05AU max
Appearance Fine white crystalline powder
Assay 99.0% min
DNase None detected
Phosphatase None detected
pKa at 25°C 6.50-7.90
Protease None detected
RNase None detected

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR